Universal Plus Roll, Brady®
Chemically inert making it ideal for use with aggressive fluids
3-ply construction makes it stronger and more durable while reducing lint. The product's green color (the universal symbol for safety) indicates that it is intended for use on hazardous materials. A dimpled roll. Perfed every 12" and up the center to help you use only what you need. For use with chemicals and hazardous fluids.
Chemically inert making it ideal for use with aggressive fluids
3-ply construction makes it stronger and more durable while reducing lint. The product's green color (the universal symbol for safety) indicates that it is intended for use on hazardous materials. A dimpled roll. Perfed every 12" and up the center to help you use only what you need. For use with chemicals and hazardous fluids.
- Catalog No:
- 10796-226
- 10796-228
- Class Type:
- Chemical
- Chemical
- Color:
- Green
- Green
Product Family Options
- DescriptionDimensionsAbsorbency
- : 107845
Specifications
Cat. No.10796-226Class TypeChemicalColorGreen - : 107848
Specifications
Cat. No.10796-228Class TypeChemicalColorGreen



