Universal Plus Pad, Brady®
Supplier: Brady Worldwide
About this item
Chemically inert making it ideal for use with aggressive fluids
3-ply construction makes it stronger and more durable while reducing lint. The product's green color (the universal symbol for safety) indicates that it is intended for use on hazardous materials. A dimpled roll. 3-ply, green, dimpled pads. For use with chemicals and hazardous fluids.
2 Options Available Below
Product Details & Documents
Chemically inert making it ideal for use with aggressive fluids
3-ply construction makes it stronger and more durable while reducing lint. The product's green color (the universal symbol for safety) indicates that it is intended for use on hazardous materials. A dimpled roll. 3-ply, green, dimpled pads. For use with chemicals and hazardous fluids.
Documents
Brady GHS Solutions Brochure
Product Literature
Specifications for Product Family
Specifications
- Catalog No:
- 10796-224
- 10796-222
- Class Type:
- Chemical
- Chemical
- Color:
- Green
- Green



