Hazwik® Absorbent Pillows, Brady®
Supplier: Brady Worldwide
Certificates
About this item
The Hazwik® Pillows feature a green spunbond outer covering to indicate their use with hazardous materials filled with green polypropylene for use with chemical and hazardous fluids.
Specifications
- Catalog No:
- 10796-280
- Class Type:
- Chemical
- Color:
- Green
Specifications
Frequently Bought Together