Hazwik® Attack Pac™ Portable Spill Kit, Brady®
Catalog # 10796-124
Supplier: Brady Worldwide
Specifications
- Description:Portable Spill Kit
- Class Type:Chemical
- Color:Silver
- Dimensions:4H x 16W x 20D"
- Cat. No.:10796-124
- Absorbency:7 gal.
Specifications
About this item
The Hazwik® Attack Pac™ Portable Spill Kit is an efficient, space saving, single use kit packed in puncture and UV resistant foil bag
Hang anywhere with the included grommet. For use with spills of hazardous materials.
Ordering information: Contents include (15) - 15" x 19" Pads, (3) - 3" x 4' SOCs, (1) - Pair Nitrile Gloves, (1) - Disposal Bag, Goggles, Instruction Sheet.