Order Entry
United States
ContactUsLinkComponent
Leupeptin
Leupeptin
  N-1000.0005BA
 :  
Leupeptin
  N-1000.0005BA
 :  4013467.0005
 :  

 

  • CAS No.:
    55123-66-5
  • Size:
    5 mg
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00037012
  • Protein/Peptide Name:
    Leupeptin
  • Molecular Weight:
    951.2
  • Sequence:
    Ac-Leu-Leu-Arg-aldehyde · sulfate salt
  • Molecular Formula:
    (C₂₀H₃₈N₆O₄)₂ · H₂SO₄
  • Cat. No.:
    N-1000.0005BA
  • Supplier no.:
    N-1000.0005

 

 

A reversible compeptitive protease inhibitor, which was shown to inhibit cathepsins B, H, L, and S, calpain and trypsin. Ac-LLR-CHO inhibits the trypsin-like activity of the proteasome endopeptidase complex. CAS Number (hemisulfate): 103476-89-7.