Order Entry
United States
ContactUsLinkComponent
(Arg8)-Vasopressin Trifluoroacetate
(Arg8)-Vasopressin Trifluoroacetate
Catalog # H-1780.0001BA
Supplier:  Bachem Americas
CAS Number:  
(Arg8)-Vasopressin Trifluoroacetate
Catalog # H-1780.0001BA
Supplier:  Bachem Americas
Supplier Number:  4012215.0001
CAS Number:  

Specifications

  • CAS No.:
    113-79-1
  • Source:
  • Species:
  • Size:
    1 mg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00804595
  • Protein Synonyms:
    Leiormone|Argipressin|AVP|Arg⁸)-Oxytocin|(Phe³|Arginine Antidiuretic Hormone
  • Protein/Peptide Name:
    (Arg8)-Vasopressin Trifluoroacetate
  • Molecular Weight:
    1084.25
  • Sequence:
    H-Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH₂
  • Molecular Formula:
    C₄₆H₆₅N₁₅O₁₂S₂
  • Cat. No.:
    H-1780.0001BA
  • Supplier no.:
    H-1780.0001

Specifications

About this item

The major physiological roles of Arg-Vasopressin are regulation of water balance in animals (antidiuretic action) and contraction of arterioles (vasopressor action). CAS number (argipressin acetate): 129979-57-3.

For desmopressin see H-7675.