Order Entry
Puerto Rico
ContactUsLinkComponent
 
H-Leu-Gly-Oh
  G-2495.0001BA
 :  
H-Leu-Gly-Oh
  G-2495.0001BA
 :  4000131.0001
 :  

 

  • CAS No.:
    686-50-0
  • Size:
    1 g
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00065940
  • Protein/Peptide Name:
    H-Leu-Gly-Oh
  • Molecular Weight:
    188.23
  • Sequence:
    H-Leu-Gly-OH
  • Molecular Formula:
    C₈H₁₆N₂O₃
  • Cat. No.:
    G-2495.0001BA
  • Supplier no.:
    G-2495.0001

 

 

The dipeptide Leu-Gly inhibits the ubiquitin-mediated proteolysis.