H-Pro-Pro-Gln-OH Trifluoroacetate
Catalog # 10796-774
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:856170-98-4
- Size:250 mg
- Storage Conditions:–20 ± 5 °C
- Protein/Peptide Name:H-Pro-Pro-Gln-OH Trifluoroacetate
- Molecular Weight:340.38
- Sequence:H-Pro-Pro-Gln-OH
- Molecular Formula:C₁₅H₂₄N₄O₅
- Cat. No.:10796-774
Specifications
About this item
Effective catalyst for conjugate addition reactions of aldehydes to β-substituted nitroolefins.