Human Hepcidin-20 Trifluoroacetate
Supplier: Bachem Americas
Certificates
About this item
Hepcidin-20, a degradation product of hepcidin-25 found in plasma and urine, shows an increased antimicrobial activity compared to the full-length peptide, but it is not involved in iron metabolism.
Specifications
- CAS No.:342790-23-2
- Species:Human
- Storage Conditions:–20 ± 5 °C
- Protein/Peptide Name:Hepcidin-20 Trifluoroacetate
- Molecular Weight:2191.77
- Sequence:H-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr-OH
- Molecular Formula:C₈₅H₁₃₅N₂₇O₂₃S₉