Order Entry
Puerto Rico
ContactUsLinkComponent
Fmoc-(D10)Leu-Oh
Fmoc-(D10)Leu-Oh
Catalog # B-4120.0500BA
Supplier:  Bachem Americas
CAS Number:  
Fmoc-(D10)Leu-Oh
Catalog # B-4120.0500BA
Supplier:  Bachem Americas
Supplier Number:  4071075.0500
CAS Number:  

Specifications

  • CAS No.:
    1190594-22-9
  • Size:
    0.5 g
  • MDL No.:
    MFCD06658583
  • Protein/Peptide Name:
    Fmoc-(D10)Leu-Oh
  • Molecular Weight:
    363.50
  • Sequence:
    Fmoc-[D₁₀]Leu-OH
  • Molecular Formula:
    C₂₁H₁₃D₁₀NO₄
  • Cat. No.:
    B-4120.0500BA
  • Supplier no.:
    B-4120.0500

Specifications

About this item

Han et al. used Fmoc-(D₁₀)-Leu-OH for obtaining labeled Ac-WL₅, a hexapeptide forming β-sheets in membranes. The aggregation of the tagged peptide was followed by neutron scattering.