Hazwik® 20-Gallon Lab Pack, Brady®
Catalog # 10796-116
Supplier: Brady Worldwide
Specifications
- Description:Drum Spill Kit
- Class Type:Chemical
- Color:Yellow
- Dimensions:18H x 21" Dia.
- Cat. No.:10796-116
- Absorbency:17 gal.
Specifications
About this item
The Hazwik® 20 Gallon Lab Pack is a UN approved water and chemical resistant polyethylene Lab Pack container with screw top lid for small to medium response of a hazardous material
Ordering information: Contents include (12) - 15" x 19" Pads, (3) - 3" x 12' SOCs, (2) - 18" x 18" Pillows, (1) - Pair Nitrile Gloves, (3) - Disposal Bags, Goggles, Emergency Response Handbook.