Human Recombinant Pleiotrophin (from E. coli)
Supplier: Peprotech
Certificates
About this item
Pleiotrophin and Midkine are structurally related heparin-binding neurotrophic factors, whose expression is developmentally regulated. The expression pattern of these neurotrophic factors suggests function in neurogenesis, cell migration, secondary organogenetic induction, and mesoderm epithelial interaction. The expression of PTN increases during the process of brain embryogenesis, and reaches maximum levels at time of birth. The physiological roles of PTN and Midkine are largely unknown, but these neurotrophins have been implicated in the pathogenesis of neuroblastomas. Recombinant Human Pleiotrophin is a 15.4 kDa protein containing 136 amino acid residues and five intra-molecular disulfide bonds.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:5764 P21246
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:Heparin Affin Regulatory Protein (HARP)|Heparin-Binding Growth Factor-8 (HBGF-8)|Osteoblast-Specific Factor 1 (OSF-1)|PTN
- Protein/Peptide Name:Pleiotrophin
- Purity:0.98
- Molecular Weight:15.4 kDa
- Sequence:GKKEKPEKKV KKSDCGEWQW SVCVPTSGDC GLGTREGTRT GAECKQTMKT QRCKIPCNWK KQFGAECKYQ FQAWGECDLN TALKTRTGSL KRALHNAECQ KTVTISKPCG KLTKPKPQAE SKKKKKEGKK QEKMLD
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together