Human Recombinant IL-36gamma (from E. coli)
Supplier: Peprotech
Certificates
About this item
The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-36 α (IL-1F6), β (IL-1F8) and γ (IL-1F9). IL-36γ is highly expressed in psoriatic plaques and in tissues containing epithelial cells. IL-36γ signals through the IL-1Rrp2 (IL-1R6) receptor, which is primarily expressed on certain dendritic cells. The interaction of the IL-1Rrp2 receptor with IL-36 ligands induces dendritic cell maturation and activation. IL-36γ also functions as an agonist of NF-κB, and can stimulate the inflammatory response in bronchial epithelial cells. Recombinant Human IL-36γ (IL-1F9) is a 17.0 kDa protein containing 152 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:56300 Q9NZH8
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:IL-1ε (epsilon) and IL-1H1|Interleukin-36γ|IL-1F9
- Protein/Peptide Name:IL-36gamma
- Purity:0.95
- Molecular Weight:17 kDa
- Sequence:SMCKPITGTI NDLNQQVWTL QGQNLVAVPR SDSVTPVTVA VITCKYPEAL EQGRGDPIYL GIQNPEMCLY CEKVGEQPTL QLKEQKIMDL YGQPEPVKPF LFYRAKTGRT STLESVAFPD WFIASSKRDQ PIILTSELGK SYNTAFELNI ND
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient