Human Recombinant IL-36beta (from E. coli)
The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-36α (IL-1F6), β (IL-1F8) and γ (IL-1F9). IL-36β is highly expressed in psoriatic plaques, and at lower levels in various other tissues. IL-36β signals through the IL-1Rrp2 (IL-1R6) receptor, which is primarily expressed on certain dendritic cells. The interaction of the IL-1Rrp2 receptor with IL-36 ligands induces dendritic cell maturation and activation. IL-36β also functions as an agonist of NF-κB, and stimulates the production of pro-inflammatory proteins, including IL-6, IL-8, BD-2, and BD-3. Recombinant Human IL-36β (IL-1F8) is a 17.3 kDa protein containing 154 amino acid residues.
: For research use only. Not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:27177 Q9NZH7
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:IL-1H2|IL-1F8|Interleukin-36β|IL-1 eta
- Protein/Peptide Name:IL-36beta
- Purity:0.98
- Molecular Weight:17.3 kDa
- Sequence:MREAAPKSYA IRDSRQMVWV LSGNSLIAAP LSRSIKPVTL HLIACRDTEF SDKEKGNMVY LGIKGKDLCL FCAEIQGKPT LQLKEKNIMD LYVEKKAQKP FLFFHNKEGS TSVFQSVSYP GWFIATSTTS GQPIFLTKER GITNNTNFYL DSVE
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient