Human Recombinant IL-1beta (from E. coli)
Supplier: Peprotech
Certificates
About this item
IL-1β is a proinflammatory cytokine produced in a variety of cells, including monocytes, tissue macrophages, keratinocytes, and other epithelial cells. Both IL-1α and IL-1β bind to the same receptor and have similar, if not identical, biological properties. These cytokines have a broad range of activities including the stimulation of thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity, and the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1α is predominantly a cell-associated cytokine. Recombinant Human IL-1β is a 17.3 kDa protein containing 153 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:3553 P01584
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:Catabolin|Lymphocyte-Activating Factor (LAF)|Endogenous Pyrogen (EP)|Interleukin-1β|Mononuclear Cell Factor (MCF)|Leukocyte Endogenous Mediator (LEM)
- Protein/Peptide Name:IL-1beta
- Purity:0.98
- Molecular Weight:17.3 kDa
- Sequence:APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient