Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant MIA-2 (from E. coli)
Human Recombinant MIA-2 (from E. coli)
Supplier:  Peprotech
Certificates
Human Recombinant MIA-2 (from E. coli)
Supplier:  Peprotech

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin Content:
    >0.1 ng/μg
  • Biological Activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    117153 Q96PC5
  • Reconstitution Instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein Synonyms:
    Melanoma Inhibitory Activity-2
  • Protein/Peptide Name:
    MIA-2
  • Purity:
    0.98
  • Molecular Weight:
    11.5 kDa
  • Sequence:
    MLESTKLLAD LKKCGDLECE ALINRVSAMR DYRGPDCRYL NFTKGEEISV YVKLAGERED LWAGSKGKEF GYFPRDAVQI EEVFISEEIQ MSTKESDFLC L
  • Endotoxin Level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping Temperature:
    Ambient

Specifications

Frequently Bought Together