Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse;Rat Cytokine
Mouse;Rat Cytokine
Catalog # 10778-858
Supplier:  Peprotech
CAS Number:  
Mouse;Rat Cytokine
Catalog # 10778-858
Supplier:  Peprotech
Supplier Number:  MHSC3
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Species:
    Rat
    Mouse
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.1 ng/μg of protein (<1 EU/μg).
  • Protein/Peptide Name:
    Cytokine
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    RT
  • Cat. No.:
    10778-858
  • Origin:
    US
  • Supplier no.:
    MHSC3

Specifications

About this item

The Murine Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo mouse hematopoietic stem cells.

The package contains the following 4 vials:
100ug Murine Flt3 Ligand
100ug Murine SCF
100ug Murine TPO
10ug Murine IL-3

To request a CoA or MSDS for the components of the package, please Contact Us.

To request a CoA or MSDS for the components of the package, please Contact Us.

Carrier free.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.