Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant M-CSF (from E. coli)
Human Recombinant M-CSF (from E. coli)
Supplier:  Peprotech
Certificates
Human Recombinant M-CSF (from E. coli)
Supplier:  Peprotech

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <0.1 ng/μg of protein (<1 EU/μg).
  • Biological Activity:
    The ED50 was determined by the dose-dependent stimulation of the proliferation of murine M-NFS-60 cells is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
  • Animal-Free:
    Y
  • Protein/Peptide Name:
    MCSF
  • Purity:
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    RT
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Form:
    Lyophilized
  • Gene ID:
    1435 P09603
  • Reconstitution Instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein Synonyms:
    MGI-IM|CSF-1|Macrophage Colony Stimulating Factor
  • Molecular Weight:
    36.8 kDa
  • Sequence:
    MEEVSEYCSH MIGSGHLQSL QRLIDSQMET SCQITFEFVD QEQLKDPVCY LKKAFLLVQD IMEDTMRFRD NTPNAIAIVQ LQELSLRLKS CFTKDYEEHD KACVRTFYET PLQLLEKVKN VFNETKNLLD KDWNIFSKNC NNSFAECSSQ GHERQSEGS
  • Endotoxin Level:
    Low

Specifications

Frequently Bought Together