Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant MIP1A (from E. coli)
Human Recombinant MIP1A (from E. coli)
Supplier:  Peprotech
Certificates
Human Recombinant MIP1A (from E. coli)
Supplier:  Peprotech

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <0.1 ng/μg of protein (<1 EU/μg).
  • Biological Activity:
    Determined by its ability to chemoattract human monocytes using a concentration range of 1.0-10.0 ng/ml.
  • Gene ID:
    6348
  • Animal-Free:
    Y
  • Protein Synonyms:
    CCL3
  • Protein/Peptide Name:
    MIP1A
  • Purity:
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together