Human Recombinant RANTES (from E. coli)
About this item
RANTES is a CC chemokine that can signal through the CCR1, CCR3, CCR5 and US28 (cytomegalovirus receptor) receptors. It is a chemoattractant towards monocytes, memory T cells (CD4+/CD45RO), basophils, and eosinophils. RANTES also has the capability to inhibit certain strains of HIV-1, HIV-2 and simian immunodeficiency virus (SIV). Recombinant Human RANTES is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
: For research use only. Not for use in diagnostic or therapeutic procedures.
6 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
RANTES is a CC chemokine that can signal through the CCR1, CCR3, CCR5 and US28 (cytomegalovirus receptor) receptors. It is a chemoattractant towards monocytes, memory T cells (CD4+/CD45RO), basophils, and eosinophils. RANTES also has the capability to inhibit certain strains of HIV-1, HIV-2 and simian immunodeficiency virus (SIV). Recombinant Human RANTES is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10778-600
- 10778-602
- 10778-604
- 10778-606
- 10778-608
- 10778-610
- Protein/Peptide Name:
- RANTES
- RANTES
- RANTES
- RANTES
- RANTES
- RANTES
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Animal-Free:
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin Content:
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- Biological Activity:
- Determined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.
- Determined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.
- Determined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.
- Determined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.
- Determined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.
- Determined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.
- Purity:
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Formulation:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Storage Conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping Temperature:
- RT
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- SizeOriginAvailabilityPrice
- Supplier Number: AF-300-06-5UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-600Protein/Peptide NameRANTESProtein/Peptide TypeRecombinantSpeciesHumanSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityDetermined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-300-06-20UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-602Protein/Peptide NameRANTESProtein/Peptide TypeRecombinantSpeciesHumanSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityDetermined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-300-06-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-604Protein/Peptide NameRANTESProtein/Peptide TypeRecombinantSpeciesHumanSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityDetermined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-300-06-250UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-606Protein/Peptide NameRANTESProtein/Peptide TypeRecombinantSpeciesHumanSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityDetermined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-300-06-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-608Protein/Peptide NameRANTESProtein/Peptide TypeRecombinantSpeciesHumanSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityDetermined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-300-06-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-610Protein/Peptide NameRANTESProtein/Peptide TypeRecombinantSpeciesHumanSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityDetermined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/ml.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT



