Human Recombinant MCP1 (from E. coli)
Supplier: Peprotech
Certificates
About this item
The MCP proteins belong to the CC chemokine family, and signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross reacts across species. Recombinant human MCP-1 is an 8.6 kDa protein containing 76 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
QPDAINAPVT CCYNFTNRKI SVQRLASYRR ITSSKCPKEA VIFKTIVAKE ICADPKQKWV QDSMDHLDKQ TQTPKT
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:−20 °C
- Endotoxin Content:<0.1 ng/μg of protein (<1 EU/μg).
- Biological Activity:Determined by its ability to chemoattract human monocytes using a concentration range of 10.0-100.0 ng/ml.
- Animal-Free:Y
- Protein Synonyms:MCAF
- Protein/Peptide Name:MCP1
- Purity:Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Formulation:Lyophilized
- Shipping Temperature:RT
Specifications
Frequently Bought Together