About this item
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and also by mast cells and NK cells. Targeted deletion of IL-13 in mice resulted in impaired Th2 cell development and indicated an important role for IL-13 in the expulsion of gastrointestinal parasites. IL-13 exerts anti-inflammatory effects on monocytes and macrophages and it inhibits the expression of inflammatory cytokines such as IL-1β, TNF-α, IL-6 and IL-8. IL-13 has also been shown to enhance B cell proliferation and to induce isotype switching, resulting in increased production of IgE. Blocking of IL-13 activity inhibits the pathophysiology of asthma. Human and murine IL-13 are cross-species reactive. Recombinant Human IL-13 is a 12.6 kDa protein consisting of 115 amino acid residues.
- Animal-free
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:3596 P35225
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:NC30|Interleukin-13
- Protein/Peptide Name:IL-13
- Purity:0.98
- Molecular Weight:12.6 kDa
- Sequence:MSPGPVPPST ALRELIEELV NITQNQKAPL CNGSMVWSIN LTAGMYCAAL ESLINVSGCS AIEKTQRMLS GFCPHKVSAG QFSSLHVRDT KIEVAQFVKD LLLHLKKLFR EGRFN`
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient