Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant Vitronectin (from HEK293 Cells)
 :  Peprotech
 
Human Recombinant Vitronectin (from HEK293 Cells)
 :  Peprotech

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <0.1 ng/μg of protein (<1 EU/μg).
  • Biological Activity:
    Testing in Progress.
  • Animal-Free:
    Y
  • Protein/Peptide Name:
    Vitronectin
  • Purity:
    Greater than 95% by SDS-PAGE gel and HPLC analyses.
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    RT