Neuregulin/Heregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3 and NRG4). To date, there are over 14 soluble and transmembrane proteins derived from the NRG1 gene. Proteolytic processing of the extracellular domain of the transmembrane NRG1 isoforms releases soluble growth factors. HRG1-β1 contains an Ig domain and an EGF-like domain; the latter is necessary for direct binding to receptor tyrosine kinases erb3 and erb4. This binding induces erb3 and erb4 heterodimerization with erb2, stimulating intrinsic kinase activity that leads to tyrosine phosphorylation. Although HRG1-β1's biological effects are still unclear, it has been found to promote motility and invasiveness of breast cancer cells, which may also involve up-regulation of expression and function of the autocrine motility-promoting factor (AMF). Recombinant Human Heregulinβ-1 (HRG1-B1) is a 7.5 kDa polypeptide consisting of only the EGF domain of heregulinβ-1 (65 amino acid residues). Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:−20 °C
- Endotoxin Content:<0.1 ng/μg of protein (<1 EU/μg).
- Biological Activity:The ED50 was determined by the dose-dependent stimulation of the proliferation of human MCF-7 cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 106 units/mg.
- Animal-Free:Y
- Protein/Peptide Name:Heregulin Beta1
- Purity:Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Formulation:Lyophilized
- Shipping Temperature:RT