Mouse Recombinant Leptin (from E. coli)
Encoded by the ob (obese) gene, Leptin is an adipose-derived cytokine that suppresses appetite and increases thermogenesis. Leptin exerts its anorectic effect via signaling through a hypothalamic receptor termed OB-R. Leptin has been shown to reduce body weight, food consumption, and plasma glucose levels in various Recombinant Murine Leptin is a 16.2 kDa protein containing 147 amino acid residues.
: For research use only. Not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:16846 P41160
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:Obesity protein (OB)
- Protein/Peptide Name:Leptin
- Purity:0.98
- Molecular Weight:16.2 kDa
- Sequence:MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSAKQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQVLTSLP SQNVLQIAND LENLRDLLHL LAFSKSCSLP QTSGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDVSPEC
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient
Frequently Bought Together