About this item
Both MIP-1α and MIP-1β are structurally and functionally related CC chemokines. They participate in host response to invading bacterial, viral, parasite and fungal pathogens, by regulating the trafficking, and activation state, of selected subgroups of inflammatory cells (e.g. macrophages, lymphocytes and NK cells). While both MIP-1α and MIP-1β exert similar effects on monocytes, their effect on lymphocytes differ; with MIP-1α selectively attracting CD8+ lymphocytes, and MIP-1β selectively attracting CD4+ lymphocytes. Additionally, MIP-1α and MIP-1β have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. Both human and murine MIP-1α and MIP-1β are active on human and murine hematopoietic cells. Recombinant Rat MIP-1α (CCL3) is a 7.8 kDa protein containing 69 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Rat
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:25542 P50229
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:Macrophage Inflammatory Protein-1α|LD78α
- Protein/Peptide Name:MIP-1alpha
- Purity:0.98
- Molecular Weight:7.8 kDa
- Sequence:APYGADTPTA CCFSYGRQIP RKFIADYFET SSLCSQPGVI FLTKRNRQIC ADPKETWVQE YITELELNA
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient