Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant IGF-BP7 (from E. coli)
 :  Peprotech
 
Human Recombinant IGF-BP7 (from E. coli)
 :  Peprotech

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin Content:
    >0.1 ng/μg
  • Biological Activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    3490 Q16270
  • Reconstitution Instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein Synonyms:
    IBP-7|Mac25|IGF binding protein-related protein-1 (IGFBP-rP1)|Insulin-like Growth Factor-Binding Protein 7
  • Protein/Peptide Name:
    IGF-BP7
  • Purity:
    0.98
  • Molecular Weight:
    26.4 kDa
  • Sequence:
    SSSDTCGPCE PASCPPLPPL GCLLGETRDA CGCCPMCARG EGEPCGGGGA GRGYCAPGME CVKSRKRRKG KAGAAAGGPG VSGVCVCKSR YPVCGSDGTT YPSGCQLRAA SQRAESRGEK AITQVSKGTC EQGPSIVTPP KDIWNVTGAQ VYLSCEVIGI PTPVLIWNKV KRGHYGVQRT ELLPGDRDNL AIQTRGGPEK HEVTGWVLVS PLSKEDAGEY ECHASNSQGQ ASASAKITVV DALHEIPVKK GEGAEL
  • Endotoxin Level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping Temperature:
    Ambient

 

Frequently Bought Together