Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant sTRAIL (from E. coli)
Human Recombinant sTRAIL (from E. coli)
Supplier:  Peprotech
Certificates
Human Recombinant sTRAIL (from E. coli)
Supplier:  Peprotech

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin Content:
    >0.1 ng/μg
  • Biological Activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    8795 O14763
  • Reconstitution Instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein Synonyms:
    CD262|soluble TRAIL Receptor-2|KILLER|TRICK2A|TRICKB|TNFRSF10B|DR5
  • Protein/Peptide Name:
    sTRAIL
  • Purity:
    0.98
  • Molecular Weight:
    14.9 kDa
  • Sequence:
    MESALITQQD LAPQQRVAPQ QKRSSPSEGL CPPGHHISED GRDCISCKYG QDYSTHWNDL LFCLRCTRCD SGEVELSPCT TTRNTVCQCE EGTFREEDSP EMCRKCRTGC PRGMVKVGDC TPWSDIECVH KES
  • Endotoxin Level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping Temperature:
    Ambient

Specifications