Human Recombinant MIP-4 (from E. coli)
MIP-4 is a CC chemokine that is expressed in lymph nodes, lungs, placenta and bone marrow. MIP-4’s primary receptor is unknown. MIP-4 chemoattracts lymphocytes, and has been shown to exert activity on both CD4+ and CD8+ T cells. Recombinant Human MIP-4 (CCL18) is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
: For research use only. Not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:6362 P55774
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:DC-CK1|AMAC-1|Macrophage Inflammatory Protein-4|PARC
- Protein/Peptide Name:MIP-4
- Purity:0.98
- Molecular Weight:7.8 kDa
- Sequence:QVGTNKELCC LVYTSWQIPQ KFIVDYSETS PQCPKPGVIL LTKRGRQICA DPNKKWVQKY ISDLKLNA
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient
Frequently Bought Together