Human Recombinant IL-12P80 (from (BTI-Tn-5B1-4) Hi-5 Insect cells)
Supplier: Peprotech
Certificates
About this item
IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Accumulating data indicates that p40 secretion precedes that of IL-12 expression. In addition, to its ability to covalently bind to p35 to form IL-12, p40 can bind to p19 to form IL-23, or it can form the homodimer designated IL-12 p80. Elevated levels of IL-12 p80 correlate to macrophage recruitment and increased inflammation in asthma and respiratory viral infection models. Recombinant Human IL-12 p80 is an 80.0 kDa disulfide linked homodimer consisting of two p40 chains of IL-12.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
- Protein/Peptide Type:Recombinant
- Source:(BTI-Tn-5B1-4) Hi-5 Insect cells
- Species:Human
- Storage Conditions:−20 °C
- Endotoxin Content:<0.1 ng/μg of protein (<1 EU/μg).
- Biological Activity:Not available.
- Protein/Peptide Name:IL-12P80
- Purity:Greater than 95% by SDS-PAGE gel and HPLC analyses.
- Formulation:Lyophilized
- Shipping Temperature:RT
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Form:Lyophilized
- Gene ID:3593 P29460
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:NKSF|CLMF|Interleukin-12
- Molecular Weight:80 kDa
- Sequence:p40 Subunit:IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS
- Endotoxin Level:Low