About this item
Thrombomodulin (TM, CD141, THBD) is an endothelial cell-expressed, transmembrane glycoprotein that can form a complex with the coagulation factor, thrombin. The thrombomodulin/thrombin complex converts protein C to its activated form, protein Ca, which in turn proteolytically cleaves and deactivates factor Va and factor VIIIa, two essential components of the coagulation mechanism. This inactivation reduces the generation of additional thrombin, and thereby effectively prevents continued coagulation. Reduced levels of thrombomodulin can correlate with the pathogenesis of certain cardiovascular diseases, such as atherosclerosis and thrombosis. However, the serum levels of the truncated circulating form of thrombomodulin are typically elevated during inflammation and in the presence of various inflammatory-related diseases. The thrombomodulin protein contains 575 amino acids, including an 18 a.a. signal sequence, a 497 a.a. extracellular domain, a 24 a.a. transmembrane sequence, and a 36 a.a. cytoplasmic region. Recombinant Human Thrombomodulin is a 51.4 kDa, 491-amino-acid length glycoprotein containing the extracellular domain of thrombomodulin.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:7056 P07204
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:CD141|THBD|TM|BDCA-3
- Protein/Peptide Name:Thrombomodulin
- Purity:0.98
- Molecular Weight:51.4 kDa
- Sequence:APAEPQPGGS QCVEHDCFAL YPGPATFLNA SQICDGLRGH LMTVRSSVAA DVISLLLNGD GGVGRRRLWI GLQLPPGCGD PKRLGPLRGF QWVTGDNNTS YSRWARLDLN GAPLCGPLCV AVSAAEATVP SEPIWEEQQC EVKADGFLCE FHFPATCRPL AVEPGAAAAA VSITYGTPFA ARGADFQALP VGSSAAVAPL GLQLMCTAPP GAVQGHWARE APGAWDCSVE NGGCEHACNA IPGAPRCQCP AGAALQADGR SCTASATQSC NDLCEHFCVP NPDQPGSYSC MCETGYRLAA DQHRCEDVDD CILEPSPCPQ RCVNTQGGFE CHCYPNYDLV DGECVEPVDP CFRANCEYQC QPLNQTSYLC VCAEGFAPIP HEPHRCQMFC NQTACPADCD PNTQASCECP EGYILDDGFI CTDIDECENG GFCSGVCHNL PGTFECICGP DSALARHIGT DCDSGKVDGG DSGSGEPPPS PTPGSTLTPP A
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient