PDGFs are disulfide-linked dimers consisting of two 12.0-13.5 kDa polypeptide chains, designated PDGF-A and PDGF-B chains. The three naturally occurring PDGFs, PDGF-AA, PDGF-BB and PDGF-AB, are potent mitogens for a variety of cell types, including smooth muscle cells, connective tissue cells, bone and cartilage cells, and some blood cells. The PDGFs are stored in platelet α-granules, and are released upon platelet activation. The PDGFs are involved in a number of biological processes, including hyperplasia, chemotaxis, embryonic neuron development, and respiratory tubule epithelial cell development. Two distinct signaling receptors used by PDGFs have been identified and named PDGFR-α and PDGFR-β. PDGFR-α is a high-affinity receptor for each of the three PDGF forms. On the other hand, PDGFR-β interacts with only PDGF-BB and PDGF-AB. Recombinant Human PDGF-BB is a 24.3 kDa disulfide-linked homodimer of two β chains (218 total amino acids).
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:5155 P01127
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:Platelet-Derived Growth Factor-BB|Glioma-Derived Growth Factor (GDGF)|Osteosarcoma-Derived Growth Factor (ODGF)
- Protein/Peptide Name:PDGF-BB
- Purity:0.98
- Molecular Weight:24.3 kDa
- Sequence:SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPIFKKATVT LEDHLACKCE TVAAARPVT
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient