Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant sCD30 (from CHO Cells)
Human Recombinant sCD30 (from CHO Cells)
Supplier:  Peprotech
Certificates
Human Recombinant sCD30 (from CHO Cells)
Supplier:  Peprotech

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage Conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin Content:
    >0.1 ng/μg
  • Biological Activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    944 P32971
  • Reconstitution Instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein Synonyms:
    TNFSF8|soluble CD30 Ligand|CD153|CD30L
  • Protein/Peptide Name:
    sCD30
  • Purity:
    0.95
  • Molecular Weight:
    21.3 kDa
  • Sequence:
    HHHHHHHHPS PGGSGGQRTD SIPNSPDNVP LKGGNCSEDL LCILKRAPFK KSWAYLQVAK HLNKTKLSWN KDGILHGVRY QDGNLVIQFP GLYFIICQLQ FLVQCPNNSV DLKLELLINK HIKKQALVTV CESGMQTKHV YQNLSQFLLD YLQVNTTISV NVDTFQYIDT STFPLENVLS IFLYSNSD
  • Endotoxin Level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping Temperature:
    Ambient

Specifications