Hydrophilic Membrane Filters, PTFE, Advantec
Advantec Hydrophilic PTFE membrane filters are ideal for HPLC and other mixtures of aqueous and organic solvents.
This non-supported membrane is very thin and optically clear when wet. Thermostable up to 100 °C.
PTFE offers maximum chemical and pH resistance to strong acids and alkalis. High flow rates with minimal aqueous extractables (<0.3 wt%).
: Note: Allowing the filter to dry will render it hydrophobic.
- Catalog No:
- 101104-798
- 101104-144
- 101103-680
- 101103-446
- 101103-456
- 101103-466
- 101104-142
- 101103-678
- 101104-784
- 101103-448
- 101103-412
- 101104-762
- 101103-416
- 101103-414
- 101104-140
- 101103-676
- 101104-290
- 101103-460
- 101103-458
- 101104-808
- 101103-462
- 101103-464
- 101103-444
- 101104-820
- Thickness:
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm