Anti-SPTLC2 Rabbit Polyclonal Antibody
Catalog # 10665-778
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Serine Palmitoyltransferase, Long Chain Base Subunit 2
- Antigen Symbol:SPTLC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:liquid
- Gene ID:9517
- Antigen Synonyms:KIAA0526|LCB2A|SPT2|LCB 2|Serine palmitoyltransferase 2|HSN1C|Long chain base biosynthesis protein 2|NSAN1C|LCB2|hLCB2a
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10665-778
- Supplier no.:BS-9027R
Specifications
About this item
This gene encodes a long chain base subunit of serine palmitoyltransferase. Serine palmitoyltransferase, which consists of two different subunits, is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine. Mutations in this gene were identified in patients with hereditary sensory neuropathy type I. [provided by RefSeq, Mar 2011].
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: SPTLC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat