Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SLU7 Rabbit Polyclonal Antibody
Anti-SLU7 Rabbit Polyclonal Antibody
Catalog # 10481-730
Supplier:  Bioss
Anti-SLU7 Rabbit Polyclonal Antibody
Catalog # 10481-730
Supplier:  Bioss
Supplier Number:  BS-8327R

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    SLU7 Homolog, Splicing Factor
  • Antigen Symbol:
    SLU7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    10569
  • Antigen Synonyms:
    hSlu7|SLU7 splicing factor homolog S. cerevisiae|9G8
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
  • Storage Temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10481-730
  • Supplier no.:
    BS-8327R

Specifications

About this item

Participates in the second catalytic step of pre-mRNA splicing, when the free hydroxyl group of exon I attacks the 3'-splice site to generate spliced mRNA and the excised lariat intron. Required for holding exon 1 properly in the spliceosome and for correct AG identification when more than one possible AG exists in 3'-splicing site region. May be involved in the activation of proximal AG. Probably also involved in alternative splicing regulation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLU7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat