Specifications
- Antibody Type:Primary
- Antigen Name:Membrane Palmitoylated Protein 6
- Antigen Symbol:MPP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:liquid
- Gene ID:51678
- Antigen Synonyms:VELI associated MAGUK 1|Palmitoylated 6 MAGUK p55 subfamily member 6|Protein associated with Lin7 2|MPP6|MAGUK protein p55T|PALS2|MPP6_HUMAN.|MAGUK p55 subfamily member 6 VELI associated MAGUK 1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10422-804
- Supplier no.:BS-5756R
Specifications
About this item
Progression of cells from interphase to mitosis involves alterations in cell structures and activities. The transition from G2 to M phase is induced by M phase promoting factor, or MPF. In M phase, many proteins are phosphorylated directly by MPF or indirectly by kinases activated by MPF. These M phase phosphoproteins (MPPs, or MPHOSPHs) permit disassembly of interphase structures and generation of M phase enzymatic activities and structures. VAM1 or MPP6 is thought to be a nucleolus specific exosome co factor, required for its role in the maturation of 5.8S rRNA.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: MPP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat