Anti-ACOX1 Rabbit Polyclonal Antibody
10406-536
: Bioss
- Antibody Type:Primary
- Antigen Name:Acyl-CoA Oxidase 1, palmitoyl
- Antigen Symbol:ACOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:liquid
- Gene ID:51
- Antigen Synonyms:Palmitoyl-CoA oxidase|Acyl-coenzyme A oxidase 1|peroxisomal|AOX
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10406-536
- Supplier no.:BS-5021R
Defects in ACOX1 are the cause of adrenoleukodystrophy pseudoneonatal (Pseudo-NALD); also known as peroxisomal acyl-CoA oxidase deficiency. Pseudo-NALD is a peroxisomal single-enzyme disorder. Clinical features include mental retardation, leukodystrophy, seizures, mild hepatomegaly, hearing deficit. Pseudo-NALD is characterized by increased plasma levels of very-long chain fatty cids, due to decreased or absent peroxisome acyl-CoA oxidase activity. Peroxisomes are intact and functioning.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: ACOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat