Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-SRSF10 Rabbit Polyclonal Antibody
  10292-142
 :  Bioss
undefined
Anti-SRSF10 Rabbit Polyclonal Antibody
  10292-142
 :  Bioss
 :  BS-13229R

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Serine/Arginine-Rich Splicing Factor 10
  • Antigen Symbol:
    SRSF10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    10772
  • Antigen Synonyms:
    TASR|SFRS13A|TASR1|SFRS13|PPP1R149|SRrp40|FUSIP1|NSSR|SRp38|TASR2|FUSIP2
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
  • Storage Temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10292-142
  • Supplier no.:
    BS-13229R

 

 

FUSIP1 is a member of the Serine/Arginine (SR) family of splicing factors. Members of the SR family all contain one or more RNA recognition motifs (RRM) and an SR-rich domain. SR factors are not only essential for constitutive splicing but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. Expressed in a variety of tissues with low expression in kidney, liver and heart, FUSIP1 localizes to the cytoplasm and nuclear speckles. In its dephosphorylated form (occurring during M phase of the cell cycle), FUSIP1 functions as a potent general repressor of pre-mRNA splicing and can interact with U1 SnRNP 70. In its phosphorylated form, FUSIP1 interacts with Tra-2∫ and, together, they may cooperate in the regulation of splicing. Four isoforms exist for FUSIP1. In neurons, FUSIP1 isoforms may act to either positively or negatively regulate alternative splicing.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SRSF10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat