Anti-HATH6 Rabbit Polyclonal Antibody
Catalog # 10268-448
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Protein atonal homolog 8
- Antigen Symbol:HATH6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:HATH-6|Helix-loop-helix protein hATH-6|ATOH8_HUMAN|Atonal homolog 8|4933425C05Rik|Atonal homolog 6|Basic helix loop helix transcription factor 6|Math6|FLJ38730|atonal homolog 8 Drosophila|Class A basic helix-loop-helix protein 21|Protein atonal homolog 8|FLJ14708|RGD1561512.|bHLHa21|Helix loop helix protein mATH-6|HATH 6|ATOH8|Okadin|hATH6
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10268-448
- Supplier no.:BS-11937R
Specifications
About this item
Putative transcription factor. May be implicated in specification and differentiation of neuronal cell lineages in the brain. May participate in kidney development and may be involved in podocyte differentiation.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: HATH6/Protein atonal homolog 8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat