Anti-CREB1 Mouse Polyclonal Antibody
100 µg
10229-772
: Bioss
: BS-0035M
Phosphorylation-dependent transcription factor that stimulates transcription upon binding to the DNA cAMP response element (CRE), a sequence present in many viral and cellular promoters. Transcription activation is enhanced by the TORC coactivators which act independently of Ser-133 phosphorylation. Involved in different cellular processes including the synchronization of circadian rhythmicity and the differentiation of adipose cells.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
- Size:100 µg
- Cross Adsorption:No
- Clonality:Polyclonal
- Purification:Purified by Protein A
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Isotype:IgG
- Host:Mouse
- Concentration:1 μg/μl
- Cat. No.:10229-772
- Reactivity:Human|Rat
- Western Blot:Yes
- Shipping Temperature:4°C
- Antibody Type:Primary
- Antigen Symbol:CREB1
- Gene ID:1385
- Immunogen:110-160/341
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:cAMP-responsive element-binding protein 1|CREB-1|CREB|CREB1|Cyclic AMP-responsive element-binding protein 1
- Antigen Name:Cyclic AMP-responsive element-binding protein 1
- Storage Temperature:Store at –20 °C for 12 months



