Anti-AHNAK2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # 10671-730
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:AHNAK nucleoprotein 2
- Antigen Symbol:AHNAK2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:113146
- Antigen Synonyms:KIAA2019|AHNAK 2|chromosome 14 open reading frame 78|C14orf78|AHNK2_HUMAN.
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10671-730
- Supplier no.:BS-9094R-CY7
Specifications
About this item
AHNAK2 contains 1 PDZ (DHR) domain. There are 3 named isoforms of AHNAK2 produced by alternative splicing. AHNAK2 is a 600-kDa protein composed of a large number of highly conserved repeat segments. AHNAKs are a class of giant propeller-like proteins that associate with calcium channel proteins of cardiomyocytes and other cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AHNAK2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat