Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CTDSPL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-CTDSPL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10480-086
Supplier:  Bioss
Anti-CTDSPL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10480-086
Supplier:  Bioss
Supplier Number:  BS-8242R-FITC

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CTD (carboxy-terminal Domain, RNA polymerase II, polypeptide A) small phosphatase like 2
  • Antigen Symbol:
    CTDSPL2
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    51496
  • Antigen Synonyms:
    CTDSP like 2|HSPC129.|CTDSP-like 2|CTD small phosphatase-like protein 2|ctdspl2|HSPC058|CTSL2_HUMAN|CTD small phosphatase like protein 2
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10480-086
  • Supplier no.:
    BS-8242R-FITC

Specifications

About this item

CTDSPL2 (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase like 2), also known as HSPC058 or HSPC129, is a 466 amino acid protein that contains one FCP1 homology domain and belongs to the CTDSPL2 family. Existing as two alternatively spliced isoforms, CTDSPL2 is thought to function as a phosphatase and is encoded by a gene that maps to human chromosome 15q15.3. Encoding more than 700 genes, chromosome 15 is made up of approximately 106 million base pairs and comprises about 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTDSPL2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: