Anti-GUSB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10474-198
: Bioss
- Antibody Type:Primary
- Antigen Name:beta Glucuronidase
- Antigen Symbol:GUSB
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:2990
- Antigen Synonyms:tcag7.927|BG|FLJ39445|MPS7
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:581-616/651
- Purification:Purified by Protein A
- Cat. No.:10474-198
- Supplier no.:BS-7980R-FITC
Plays an important role in the degradation of dermatan and keratan sulfates.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: beta glucuronidase
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat