To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Vacuolar Protein Sorting 4A
- Antigen Symbol:VPS4A
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:27183
- Antigen Synonyms:VPS4|vacuolar sorting protein 4|SKD1-homolog|hVPS4|VPS4-1|SKD2|vacuolar protein sorting factor 4A|SKD1A|SKD1|vacuolar protein sorting-associated protein 4A
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10469-510
- Supplier no.:BS-7777R-FITC
Specifications
About this item
Involved in intracellular protein transport probably out of a prevacuolar endosomal compartment. May be involved in the release of components of the bilayered coat from the endosomal membrane. The association with ESCRT-III complex mediates the ATP-dependent disassembly of the ESCRT-III complex. In case of infection, the HIV-1 virus takes advantage of it for budding and exocytic cargos of viral proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: VPS4A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat