Anti-SPON1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # 10463-824
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Spondin 1
- Antigen Symbol:SPON1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:10418
- Antigen Synonyms:f-spondin|VSGP/F-spondin|MGC10724|KIAA0762
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:775-807/807
- Purification:Purified by Protein A
- Cat. No.:10463-824
- Supplier no.:BS-7520R-A555
Specifications
About this item
Cell adhesion protein that promotes the attachment of spinal cord and sensory neuron cells and the outgrowth of neurites in vitro. May contribute to the growth and guidance of axons in both the spinal cord and the PNS (By similarity). Major factor for vascular smooth muscle cell.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: SPON1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat