Order Entry
United States
ContactUsLinkComponent
Anti-S1PR1 Rabbit Polyclonal Antibody (Cy5®)
Anti-S1PR1 Rabbit Polyclonal Antibody (Cy5®)
  10461-970
 :  Bioss
Anti-S1PR1 Rabbit Polyclonal Antibody (Cy5®)
  10461-970
 :  Bioss
 :  BS-7112R-CY5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Sphingosine-1-phosphate Receptor 1
  • Antigen Symbol:
    S1PR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1901
  • Antigen Synonyms:
    CD363|CHEDG1|Sphingosine 1-phosphate receptor 1|S1P receptor 1|ECGF1|D1S3362|EDG-1|Endothelial differentiation G-protein coupled receptor 1|EDG1|S1P1
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    106-146/382
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10461-970
  • Supplier no.:
    BS-7112R-CY5

 

 

G-protein coupled receptor for the bioactive lysosphingolipid sphingosine 1-phosphate (S1P) that seems to be coupled to the G(i) subclass of heteromeric G proteins. Signaling leads to the activation of RAC1, SRC, PTK2/FAK1 and MAP kinases. Plays an important role in cell migration, probably via its role in the reorganization of the actin cytoskeleton and the formation of lamellipodia in response to stimuli that increase the activity of the sphingosine kinase SPHK1. Required for normal chemotaxis toward sphingosine 1-phosphate. Required for normal embryonic heart development and normal cardiac morphogenesis. Plays an important role in the regulation of sprouting angiogenesis and vascular maturation. Inhibits sprouting angiogenesis to prevent excessive sprouting during blood vessel development. Required for normal egress of mature T-cells from the thymus into the blood stream and into peripheral lymphoid organs. Plays a role in the migration of osteoclast precursor cells, the regulation of bone mineralization and bone homeostasis (By similarity). Plays a role in responses to oxidized 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphocholine by pulmonary endothelial cells and in the protection against ventilator-induced lung injury.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: S1PR1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat