Anti-SCN3B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10456-964
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Sodium Channel Voltage-gated Type 3 Beta
- Antigen Symbol:SCN3B
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:55800
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10456-964
- Supplier no.:BS-6881R-FITC
Specifications
About this item
SCN3B is one member of the sodium channel beta subunits of voltage-gated sodium channels, which are responsible for the generation and propagation of action potentials in neurons and muscle. SCN3B influences the inactivation kinetics of the sodium channel.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SCN3B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat