Anti-SLC26A4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # 10454-736
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 26 Member A4
- Antigen Symbol:SLC26A4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:5172
- Antigen Synonyms:TDH2B|EVA|PDS|DFNB4
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:317-344/780
- Purification:Purified by Protein A
- Cat. No.:10454-736
- Supplier no.:BS-6787R-A555
Specifications
About this item
Sodium-independent transporter of chloride and iodide.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC26A4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat