Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-KIRREL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-KIRREL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10453-328
Supplier:  Bioss
Anti-KIRREL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10453-328
Supplier:  Bioss
Supplier Number:  BS-6721R-FITC

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Kin of IRRE Like 2
  • Antigen Symbol:
    KIRREL2
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    84063
  • Antigen Synonyms:
    FILTRIN|NEPH3|NLG1
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    195-242/708
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10453-328
  • Supplier no.:
    BS-6721R-FITC

Specifications

About this item

KIRREL2 is an immunoglobulin super family gene, it has been found to be expressed in beta cells of pancreatic islets; KIRR2_HUMAN.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIRREL2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat